Ácido (2,6-dimetilfenil)metabórico ≥98.0% (by HPLC) (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: Dihidroxi(2,6-dimetilfenil)borano

D3633-1G D3633-5G
TCIAD3633-1GEA 19 EUR
TCIAD3633-1G TCIAD3633-5G
Ácido (2,6-dimetilfenil)metabórico ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
Ácido (2,6-dimetilfenil)metabórico
Formula: C₈H₁₁BO₂
MW: 149,98 g/mol
Número MDL: MFCD01009693
Núm. CAS: 100379-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bb88ba786897bac349dc842f7882623697f4790&vid=7bb88ba786897bac349dc842f7882623697f4790&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR