Ácido (3,5-dimetilfenil)metabórico (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid
D3396-25G
D3396-5G
TCIAD3396-25GEA
160
EUR
TCIAD3396-25G
TCIAD3396-5G
Ácido (3,5-dimetilfenil)metabórico (contains varying amounts of Anhydride)
Ácido (3,5-dimetilfenil)metabórico
Formula:
C₈H₁₁BO₂ MW: 149.98 g/mol |
Número MDL:
MFCD00185689 Núm. CAS: 172975-69-8 |
|
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | 97.0 to 115.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...