Sulfato de guanidinio ≥99.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Guanidine sulphate

G0166-25G G0166-500G
TCIAG0166-25GEA 20 EUR
TCIAG0166-25G TCIAG0166-500G
Sulfato de guanidinio ≥99.0% (by titrimetric analysis)
Sulfato de guanidinio
Formula: C₂H₁₂N₆O₄S
Número MDL: MFCD00013115
Núm. CAS: 594-14-9
EINECS: 209-826-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water almost transparency
Drying loss max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR