Ácido 2,7-(bis(2-arsonofenilazo))-1,8-dihidroxinaftaleno-3,6-disulfonico ≥95.0% (by titrimetric analysis) para espectrofotometría
Proveedor: TCI
|
Peligro
|
Sinónimos:
2,7-Bis(2-arsonophenylazo)chromotropic acid, 2,7-Bis(2-arsonophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic acid, 2,2′-(1,8-Dihydroxy-3,6-disulphonaphthylene-2,7-bisazo)bisbenzenearsonic acid, Arsenazo Ⅲ
A5009-1G
A5009-5G
TCIAA5009-1GEA
35
EUR
TCIAA5009-1G
TCIAA5009-5G
Ácido 2,7-(bis(2-arsonofenilazo))-1,8-dihidroxinaftaleno-3,6-disulfonico ≥95.0% (by titrimetric analysis) para espectrofotometría
Ácido 2,7-(bis(2-arsonofenilazo))-1,8-dihidroxinaftaleno-3,6-disulfonico
Spectrophotometric reagent for U, Th, Zr and other metals, Indicator for the precipitation titration of SO4 with Ba
Formula:
C₂₂H₁₈As₂N₄O₁₄S₂ MW: 776,38 g/mol |
Número MDL:
MFCD00036695 Núm. CAS: 1668-00-4 EINECS: 216-788-6 UN: 3465 ADR: 6.1,II |
Resultados test especificaciones
Appearance | Dark green to Dark red to Black powder to crystaline |
Purity (Neutralization titration) | min. 95.0 % (after drying) |
Lambda max. | 535.0 to 545.0 nm |
Absorbance | min. 0.600 (16mg/L, 535.0 to 545.0 nm) (calcd.on dried substance) |
Drying loss | max. 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...