Carbonato de bencilo y succinimido ≥98.0% (by total nitrogen basis)

Proveedor: TCI

Sinónimos: CBZ-OSu (Z-OSu)

C1124-25G C1124-250G
TCIAC1124-25GEA 24 EUR
TCIAC1124-25G TCIAC1124-250G
Carbonato de bencilo y succinimido ≥98.0% (by total nitrogen basis)
Carbonato de bencilo y succinimido
Formula: C₁₂H₁₁NO₅
MW: 249,22 g/mol
Número MDL: MFCD00005513
Núm. CAS: 13139-17-8
EINECS: 236-075-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea677ed6a44c876fd66e9c9ae32c0410972ff238&vid=ea677ed6a44c876fd66e9c9ae32c0410972ff238&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 79.0 to 83.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR