5-Nitro-1,2,3,4-tetrahidroftalazina-1,4-diona ≥97.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 2,3-Dihydro-5-nitro-1,4-phthalazinedione, 5-Nitro-2,3-dihydrophthalazine-1,4-dione, 3-Nitrophthalic Hydrazide, 3-Nitrophthalhydrazide

N0622-25G N0622-5G
TCIAN0622-25GEA 197 EUR
TCIAN0622-25G TCIAN0622-5G
5-Nitro-1,2,3,4-tetrahidroftalazina-1,4-diona ≥97.0% (by titrimetric analysis)
5-Nitro-1,2,3,4-tetrahidroftalazina-1,4-diona
Formula: C₈H₅N₃O₄
MW: 207,1430 g/mol
Número MDL: MFCD00006889
Núm. CAS: 3682-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f261031fcb8ce28e449acc4b5ebe762683f2c471&vid=f261031fcb8ce28e449acc4b5ebe762683f2c471&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR