4'-Bromo-3'-methylacetanilide ≥98.0% (por GC)

Proveedor: TCI

B3142-25G B3142-5G
TCIAB3142-25GEA 50 EUR
TCIAB3142-25G TCIAB3142-5G
4'-Bromo-3'-methylacetanilide ≥98.0% (por GC)
4'-Bromo-3'-methylacetanilide
Formula: C₉H₁₀BrNO
MW: 228,09 g/mol
Número MDL: MFCD00040822
Núm. CAS: 90914-81-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c62d8696f21aff5eff0bd8afed6a39cd93eaff3&vid=0c62d8696f21aff5eff0bd8afed6a39cd93eaff3&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR