trans-4-Pentylcyclohexanecarboxylic acid ≥99.0% (by GC, titration analysis)

Proveedor: TCI
Atención

P0944-25G P0944-5G
TCIAP0944-25GEA 99 EUR
TCIAP0944-25G TCIAP0944-5G
trans-4-Pentylcyclohexanecarboxylic acid ≥99.0% (by GC, titration analysis)
trans-4-Pentylcyclohexanecarboxylic acid
Formula: C₁₁H₂₀O₂
MW: 198,30 g/mol
Número MDL: MFCD00009964
Núm. CAS: 38289-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98f13e983c0d8a25ae0a5802e7380a504e65505d&vid=98f13e983c0d8a25ae0a5802e7380a504e65505d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 54.0 to 57.0 deg-C
Mesomorphic range 49.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR