4-Chloro-DL-phenylalanine ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
p-Chloro-DL-Phenylalanine, H-DL-Phe(4-Cl)-OH
C0253-1G
C0253-25G
TCIAC0253-1GEA
18
EUR
TCIAC0253-1G
TCIAC0253-25G
4-Chloro-DL-phenylalanine ≥98.0% (by titrimetric analysis)
4-Chloro-DL-phenylalanine
Formula:
C₉H₁₀ClNO₂ MW: 199,6 g/mol |
Número MDL:
MFCD00002601 Núm. CAS: 7424-00-2 EINECS: 231-051-9 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...