Levodopa ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa

D0600-25G D0600-5G D0600-100G
TCIAD0600-25GEA 60 EUR
TCIAD0600-25G TCIAD0600-5G TCIAD0600-100G
Levodopa ≥98.0% (by HPLC, titration analysis)
Levodopa
Formula: C₉H₁₁NO₄
Número MDL: MFCD00002598
Núm. CAS: 59-92-7
EINECS: 200-445-2
Índice Merck: 14,05464
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca0c0cac1831886d7c48a94772f95dcc4fca0784&vid=ca0c0cac1831886d7c48a94772f95dcc4fca0784&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -11.0 to -13.0 deg (C=5, 1mol/L HCl)
Solubility in 1mol/L HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR