2,8-Dibromdibenzothiophene ≥96.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
2,8-Dibromodibenzo[b,d]thiophene
D2245-1G
D2245-25G
D2245-5G
TCIAD2245-1GEA
31
EUR
TCIAD2245-1G
TCIAD2245-25G
TCIAD2245-5G
2,8-Dibromdibenzothiophene ≥96.0% (por GC)
2,8-Dibromdibenzothiophene
Formula:
C₁₂H₆Br₂S MW: 342,0490 g/mol |
Número MDL:
MFCD00092755 Núm. CAS: 31574-87-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Solubility in hot Toluene | almost transparency |
Melting point | 223.0 to 228.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...