7-Hydroxyflavanone ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
H1006-1G
H1006-5G
TCIAH1006-1GEA
50
EUR
TCIAH1006-1G
TCIAH1006-5G
7-Hydroxyflavanone ≥98.0% (by HPLC, titration analysis)
7-Hydroxyflavanone
Formula:
C₁₅H₁₂O₃ MW: 240,25 g/mol |
Número MDL:
MFCD00017487 Núm. CAS: 6515-36-2 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 187.0 to 191.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...