trans-4-Hydroxycyclohexanecarboxylic acid ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Atención

H1175-25G H1175-5G
TCIAH1175-25GEA 187 EUR
TCIAH1175-25G TCIAH1175-5G
trans-4-Hydroxycyclohexanecarboxylic acid ≥97.0% (by GC, titration analysis)
trans-4-Hydroxycyclohexanecarboxylic acid
Formula: C₇H₁₂O₃
MW: 144,1684 g/mol
Número MDL: MFCD02093477
Núm. CAS: 3685-26-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b92eb5e3224bc5fa5668397bd1e693ca48584b3&vid=5b92eb5e3224bc5fa5668397bd1e693ca48584b3&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 144.0 to 149.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR