N-Fmoc-L-leucina ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

F0295-1G F0295-25G F0295-5G
TCIAF0295-1GEA 16 EUR
TCIAF0295-1G TCIAF0295-25G TCIAF0295-5G
N-Fmoc-L-leucina ≥98.0% (by HPLC, titration analysis)
N-Fmoc-L-leucine
Formula: C₂₁H₂₃NO₄
MW: 353,4 g/mol
Número MDL: MFCD00037133
Núm. CAS: 35661-60-0
EINECS: 252-662-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d5aa9397369c0c6b589eaa3ed5ac598dcc13bbc&vid=6d5aa9397369c0c6b589eaa3ed5ac598dcc13bbc&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 154.0 to 157.0 deg-C
Specific rotation [a]20/D -24.0 to -26.0 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR