Capillary columns, OPTIMA® δ
Proveedor: MACHEREY-NAGEL
726400.25 726400.50 726410.10 726410.20 726420.30 726420.60 726421.30 726440.30 726440.60 726441.30 726441.60 726442.30 726442.60 726443.30 726465.25 726470.60 726480.60 726481.30 726481.60 726482.30 726483.30 726490.10 726482.60
MANA726400.25EA 720 EUR
MANA726400.25 MANA726400.50 MANA726410.10 MANA726410.20 MANA726420.30 MANA726420.60 MANA726421.30 MANA726440.30 MANA726440.60 MANA726441.30 MANA726441.60 MANA726442.30 MANA726442.60 MANA726443.30 MANA726465.25 MANA726470.60 MANA726480.60 MANA726481.30 MANA726481.60 MANA726482.30 MANA726483.30 MANA726490.10 MANA726482.60
Capillary columns, OPTIMA® δ
Columnas de cromatografía
Medium polar GC phases with autoselectivity.
  • Analytes determine the polarity of the phase
  • Unique from MN, no similar phase
  • Ideal for MSD and PND detectors
For columns with 0,10 to 0,32 mm ID the max. temperature for isothermal operation is 340 °C, the max. temperature for short isotherms in a temperature program is 360 °C; for 0,53 mm ID columns the max. temperatures are 320 and 340 °C, respectively.
Polysiloxane phase without CN groups (better bleeding behaviour).
OPTIMA® δ-3: Autoselective polarity between polarities 5 and 1701.
OPTIMA® δ-6: Autoselective polarity between polarities 17 and 210.