Ácido (2-hidroxifenil)metabórico (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 2-Hydroxybenzeneboronic acid, Dihydroxy(2-hydroxyphenyl)borane, (2-Hydroxyphenyl)metaboric acid

H1184-1G H1184-5G
TCIAH1184-1GEA 39 EUR
TCIAH1184-1G TCIAH1184-5G
Ácido (2-hidroxifenil)metabórico (contains varying amounts of Anhydride)
Ácido (2-hidroxifenil)metabórico
Formula: C₆H₇BO₃
MW: 137.93 g/mol
Número MDL: MFCD01074581
Núm. CAS: 89466-08-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4de7cb4e35a16fa7e2142c8e5166761d4a6fd9be&vid=4de7cb4e35a16fa7e2142c8e5166761d4a6fd9be&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 115.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR