Ácido (3-etoxifenil)metabórico (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

E0724-1G E0724-5G
TCIAE0724-1GEA 20 EUR
TCIAE0724-1G TCIAE0724-5G
Ácido (3-etoxifenil)metabórico (contains varying amounts of Anhydride)
Ácido (3-etoxifenil)metabórico
Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
Número MDL: MFCD00274219
Núm. CAS: 90555-66-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93fda71bf6617edc3ad4239d6b131d0c3350b556&vid=93fda71bf6617edc3ad4239d6b131d0c3350b556&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR