Ácido (4-nitrofenoxi)acético ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-(4-Nitrophenoxy)acetic acid, 4-Nitrophenoxyacetic acid

N0226-25G N0226-500G
TCIAN0226-25GEA 39 EUR
TCIAN0226-25G TCIAN0226-500G
Ácido (4-nitrofenoxi)acético ≥98.0% (by GC, titration analysis)
Ácido (4-nitrofenoxi)acético
Formula: (C₆H₄(NO₂))OCH₂COOH
MW: 197,15 g/mol
Número MDL: MFCD00017030
Núm. CAS: 1798-11-4
EINECS: 217-283-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ada7602283cb365130552f2bb504aee3aa4a201d&vid=ada7602283cb365130552f2bb504aee3aa4a201d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 187.0 to 190.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR