1-Butildihidroxiborano (contains varying amounts of Anhydride) for esterification
Proveedor: TCI
|
Atención
|
Sinónimos:
Butylboronic acid, n-Butylboronic acid
B0529-1G
B0529-25G
B0529-5G
TCIAB0529-1GEA
20
EUR
TCIAB0529-1G
TCIAB0529-25G
TCIAB0529-5G
1-Butildihidroxiborano (contains varying amounts of Anhydride) for esterification
1-Butildihidroxiborano
Formula:
C₄H₁₁BO₂ MW: 101.94 g/mol |
Número MDL:
MFCD00002106 Núm. CAS: 4426-47-5 EINECS: 224-607-7 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystaline |
Purity (Neutralization titration) | 97.0 to 120.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...