1-(4-(Trifluorometil)fenil)piperazina ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
N-(α,α,α-Trifluoro-p-tolyl)piperazine
T2741-25G
T2741-5G
TCIAT2741-25GEA
247
EUR
TCIAT2741-25G
TCIAT2741-5G
1-(4-(Trifluorometil)fenil)piperazina ≥98.0% (by GC, titration analysis)
1-(4-(Trifluorometil)fenil)piperazina
Formula:
C₁₁H₁₃F₃N₂ MW: 230,23 g/mol |
Número MDL:
MFCD00040765 Núm. CAS: 30459-17-7 EINECS: 250-210-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...