Ácido 2-furoico ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Furan-2-carboxylic acid, 2-Furancarboxylic acid

F0081-25G F0081-500G
TCIAF0081-25GEA 16 EUR
TCIAF0081-25G TCIAF0081-500G
Ácido 2-furoico ≥98.0% (by GC, titration analysis)
Ácido 2-furoico
Formula: C₅H₄O₃
MW: 112,08 g/mol
Número MDL: MFCD00003238
Núm. CAS: 88-14-2
EINECS: 201-803-0
Índice Merck: 14,04307
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf1d79bb506cb52aabd95c87f7e8b3413f444062&vid=bf1d79bb506cb52aabd95c87f7e8b3413f444062&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 130.0 to 133.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR