(S)-(+)-Ethyl mandelate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: L-(+)-Mandelic acid ethyl ester, (S)-Ethyl 2-hydroxy-2-phenylacetate, Ethyl L-(+)-Mandelate

M1273-25G M1273-5G
TCIAM1273-25GEA 128 EUR
TCIAM1273-25G TCIAM1273-5G
(S)-(+)-Ethyl mandelate ≥98.0% (por GC)
(S)-(+)-Ethyl mandelate
Formula: C₁₀H₁₂O₃
MW: 180,2 g/mol
Pto. Ebullición: 107°C
Número MDL: MFCD00064249
Núm. CAS: 13704-09-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d4f80f757902ecc7c17f6e0df2961a67d7d7f73&vid=5d4f80f757902ecc7c17f6e0df2961a67d7d7f73&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Freezing point 27.0 to 30.0 deg-C
Specific rotation [a]20/D +135.0 to +140.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR