L(+)-α-Fenilglicina ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
H-Phg-OH, L-(+)-2-Phenylglycine
P1288-25G
P1288-100G
TCIAP1288-25GEA
26
EUR
TCIAP1288-25G
TCIAP1288-100G
L(+)-α-Fenilglicina ≥98.0% (by HPLC, titration analysis)
L(+)-α-Fenilglicina
Formula:
C₈H₉NO₂ MW: 151,16 g/mol |
Número MDL:
MFCD00064403 Núm. CAS: 2935-35-5 EINECS: 220-909-8 Índice Merck: 14,07291 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 98.0 ee% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +156.0 to +160.0 deg (C=1, 1mol/L HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...