Dicloruro de etilendiamonio ≥98.0% (by total nitrogen basis)

Proveedor: TCI
Atención

Sinónimos: Ethylenediamine dihydrochloride

E0078-25G E0078-500G
TCIAE0078-25GEA 16 EUR
TCIAE0078-25G TCIAE0078-500G
Dicloruro de etilendiamonio ≥98.0% (by total nitrogen basis)
Dicloruro de etilendiamonio
Formula: C₂H₁₀Cl₂N₂
MW: 133,02 g/mol
Número MDL: MFCD00012524
Núm. CAS: 333-18-6
EINECS: 206-369-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49f7e5804a5a307745b33b255f86d65f9436556d&vid=49f7e5804a5a307745b33b255f86d65f9436556d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 % (calcd.on dried substance)
Solubility in Water very faint turbidity
Drying loss max. 3.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR