6-Chloro-1,3-dimethyluracil ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 6-Chloro-1,3-dimethylpyrimidine-2,4(1H,3H)-dione, 1,3-Dimethyl-6-chlorouracil

D1619-25G D1619-5G
TCIAD1619-25GEA 95 EUR
TCIAD1619-25G TCIAD1619-5G
6-Chloro-1,3-dimethyluracil ≥98.0% (by GC, titration analysis)
6-Chloro-1,3-dimethyluracil
Formula: C₆H₇ClN₂O₂
MW: 174,59 g/mol
Número MDL: MFCD00038066
Núm. CAS: 6972-27-6
EINECS: 230-205-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b75ba6ab9a44faddc17de4ac2f41344d87fc844&vid=4b75ba6ab9a44faddc17de4ac2f41344d87fc844&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 110.0 to 114.0 deg-C
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR