Glicinamida clorhidrato (H-Gly-NH2.HCl) ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: H-Gly-NH2.HCl

G0193-100G G0193-25G
TCIAG0193-100GEA 218 EUR
TCIAG0193-100G TCIAG0193-25G
Glicinamida clorhidrato (H-Gly-NH2.HCl) ≥98.0% (by titrimetric analysis)
Glicinamida clorhidrato (H-Gly-NH2.HCl)
Formula: C₂H₇ClN₂O
MW: 110,54 g/mol
Número MDL: MFCD00013008
Núm. CAS: 1668-10-6
EINECS: 216-789-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7100d4eaac98606c24c072fd3877e98fb30bcdd&vid=d7100d4eaac98606c24c072fd3877e98fb30bcdd&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR