Ácido 2-(p-clorofenoxi)-2-metilpropionico ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 2-(4-Chlorophenoxy)-2-methylpropionic acid, Clofibric acid, 2-(4-Chlorophenoxy)-2-methylpropanoic acid

C0940-25G C0940-500G
TCIAC0940-25GEA 28 EUR
TCIAC0940-25G TCIAC0940-500G
Ácido 2-(p-clorofenoxi)-2-metilpropionico ≥97.0% (by titrimetric analysis)
Ácido 2-(p-clorofenoxi)-2-metilpropionico
Formula: C₁₀H₁₁ClO₃
MW: 214,65 g/mol
Número MDL: MFCD00004192
Núm. CAS: 882-09-7
EINECS: 212-925-9
Índice Merck: 14,02378
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38b67ac14f88f316fd4a9ac3e10060f3bd6a23ed&vid=38b67ac14f88f316fd4a9ac3e10060f3bd6a23ed&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 119.0 to 122.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR