2,6-Dinitro-4-(trifluoromethyl)phenol ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

D2880-25G D2880-5G
TCIAD2880-25GEA 183 EUR
TCIAD2880-25G TCIAD2880-5G
2,6-Dinitro-4-(trifluoromethyl)phenol ≥97.0% (by GC, titration analysis)
2,6-Dinitro-4-(trifluoromethyl)phenol
Formula: C₇H₃F₃N₂O₅
Número MDL: MFCD00194260
Núm. CAS: 393-77-1
UN: 2930
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cf6ab55d348ddf82fa2c419fc9081ca3decfab0&vid=9cf6ab55d348ddf82fa2c419fc9081ca3decfab0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR