Dimethyl-2,3-pyridinedicarboxylate ≥98.0% (by GC, titration analysis)

Proveedor: TCI

P1303-25G P1303-5G
TCIAP1303-25GEA 103 EUR
TCIAP1303-25G TCIAP1303-5G
Dimethyl-2,3-pyridinedicarboxylate ≥98.0% (by GC, titration analysis)
Dimethyl-2,3-pyridinedicarboxylate
Formula: C₉H₉NO₄
Número MDL: MFCD00186549
Núm. CAS: 605-38-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e551dc20e255bd0716237fcf18eb1634a3e30e0a&vid=e551dc20e255bd0716237fcf18eb1634a3e30e0a&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 55.0 to 58.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR