D-(-)-Isoascorbic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI

A0520-25G A0520-500G
TCIAA0520-25GEA 16 EUR
TCIAA0520-25G TCIAA0520-500G
D-(-)-Isoascorbic acid ≥98.0% (by titrimetric analysis)
D-(-)-Isoascorbic acid
Formula: C₆H₈O₆
Número MDL: MFCD00005378
Núm. CAS: 89-65-6
EINECS: 201-928-0
Índice Merck: 14,05126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbd57834ee0719ce2bff91af8c97a465335486d9&vid=dbd57834ee0719ce2bff91af8c97a465335486d9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Specific rotation [a]20/D -16.0 to -19.0 deg (C=10, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR