Capillary columns, OPTIMA® WAX
Proveedor: MACHEREY-NAGEL
726296.25 726296.30 726296.50 726296.60 726321.25 726321.30 726321.50 726321.60 726548.30 726549.25 726549.30 726600.25 726600.30 726600.50 726600.60
MANA726296.25EA 630 EUR
MANA726296.25 MANA726296.30 MANA726296.50 MANA726296.60 MANA726321.25 MANA726321.30 MANA726321.50 MANA726321.60 MANA726548.30 MANA726549.25 MANA726549.30 MANA726600.25 MANA726600.30 MANA726600.50 MANA726600.60
Capillary columns, OPTIMA® WAX
Columnas de cromatografía
Polar GC phase.
  • Recommended for solvent analysis and alcohols, suitable for aqueous solutions
  • Maximum temperature for isothermal operation 240 °C (0,53 mm ID columns: 220 °C)
  • Maximum temperature for short isotherms in a temperature program 250 °C (0,53 mm ID columns: 240 °C)
Polyethylene glycol phase.
USP code: G16.