Capillary columns, OPTIMA® FFAP
Proveedor: MACHEREY-NAGEL
726116.25 726116.30 726116.50 726116.60 726341.25 726341.30 726341.50 726341.60 726344.25 726344.30 726344.50 726345.30 726180.10 726346.25
MANA726116.25EA 590 EUR
MANA726116.25 MANA726116.30 MANA726116.50 MANA726116.60 MANA726341.25 MANA726341.30 MANA726341.50 MANA726341.60 MANA726344.25 MANA726344.30 MANA726344.50 MANA726345.30 MANA726180.10 MANA726346.25
Capillary columns, OPTIMA® FFAP
Columnas de cromatografía
Polar GC phase.
  • Ideal for fatty acid methyl esters (FAMEs), free carboxylic acids
  • Maximum temperature for isothermal operation 250 °C (0,53 mm ID columns: 220 °C)
  • Maximum temperature for short isotherms in a temperature program 260 °C (0,53 mm ID columns: 240 °C)
Polyethylene glycol 2-nitroterephthalate phase.
USP code: G35 / close equivalent to G25.