Capillary columns, PERMABOND® SE-52
Proveedor: MACHEREY-NAGEL
723054.25 723310.25 723312.25
MANA723054.25EA 590 EUR
MANA723054.25 MANA723310.25 MANA723312.25
Capillary columns, PERMABOND® SE-52
Columnas de cromatografía
  • Nonpolar phase
  • Maximum temperature 300 °C (long-term temperature)
  • Maximum temperature 320 °C (short-term max. temperature in a temperature program)