QuEChERS dispersive kits
Proveedor: Agilent
5982-5456 5982-5022 5982-5058 5982-5121 5982-5122 5982-5156 5982-5158 5982-5221 5982-5222 5982-5256 5982-5258 5982-5321 5982-5021 5982-5421 5982-5356 5982-5356CH 5982-4921 5982-4956 5982-5021CH 5982-5022CH 5982-5056CH 5982-5058CH 5982-5121CH 5982-5156CH 5982-5158CH 5982-5256CH 5982-5258CH
HEWL5982-5456EA 315 EUR
HEWL5982-5456 HEWL5982-5022 HEWL5982-5058 HEWL5982-5121 HEWL5982-5122 HEWL5982-5156 HEWL5982-5158 HEWL5982-5221 HEWL5982-5222 HEWL5982-5256 HEWL5982-5258 HEWL5982-5321 HEWL5982-5021 HEWL5982-5421 HEWL5982-5356 HEWL5982-5356CH HEWL5982-4921 HEWL5982-4956 HEWL5982-5021CH HEWL5982-5022CH HEWL5982-5056CH HEWL5982-5058CH HEWL5982-5121CH HEWL5982-5156CH HEWL5982-5158CH HEWL5982-5256CH HEWL5982-5258CH
QuEChERS dispersive kits
Columnas de cromatografía
Step 2: Dispersive SPE cleanup
Select the dispersive SPE kit suited to the type of food being analysed and the method you are following. In this step, an aliquot of the sample extract from step one (see QuEChERS extraction kits) is added to a 2 or 15 ml centrifuge tube containing a small amount of SPE sorbent and MgSO₄. The sorbent will pull out interfering matrix materials from the sample, while the MgSO₄ helps remove excess water and improve analyte partitioning.