Screw caps for micro tubes
Proveedor: SARSTEDT
65.716.001 65.716.003 65.716.005 65.716.008 65.716.022 65.716.004 65.716.006 65.716.007 65.716.999 65.716
SARS65.716.001EA 53.82 EUR
SARS65.716.001 SARS65.716.003 SARS65.716.005 SARS65.716.008 SARS65.716.022 SARS65.716.004 SARS65.716.006 SARS65.716.007 SARS65.716.999 SARS65.716
Screw caps for micro tubes
Tapas Tapones para tubos de ensayo
PP, coloured.
  • Excellent chemical resistance
  • Excellent temperature resistance
Screw cap with O-ring seal.