Capillary column splitter kits
Proveedor: Agilent
5181-3390 5181-3391 5181-3392
HEWL5181-3390EA 519 EUR
HEWL5181-3390 HEWL5181-3391 HEWL5181-3392
Capillary column splitter kits
Racores y conectores
These splitters are used for splitting an injected sample from a single injector into two columns and on to two detectors.
The dimensions of both the columns, length and internal diameter (ID), should be the same so that the vaporized sample will get spilt into almost equal amount on to two columns.
Información de suministro: Kit which comes with transfer line, splitter union and necessary ferrules.