Anti-SLAMF7/CS1 Rabbit Monoclonal Antibody [clone: EPR22948-114] (APC (Allophycocyanin))
Proveedor: Abcam
AB313957-10UL AB313957-100UL
ABCAAB313957-10ULEA 0 EUR
ABCAAB313957-10UL ABCAAB313957-100UL
Anti-SLAMF7/CS1 Rabbit Monoclonal Antibody [clone: EPR22948-114] (APC (Allophycocyanin))
Anticuerpos
APC Rabbit monoclonal [EPR22948-114] to SLAMF7/CS1.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SLAMF7/CS1
Clonality: Monoclonal
Clone: EPR22948-114
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human