Anti-MASP1 Rabbit Monoclonal Antibody [clone: EPR22680-36]
Proveedor: Abcam
AB255814-100 AB255814-1ML
ABCAAB255814-100EA 757 EUR
ABCAAB255814-100 ABCAAB255814-1ML
Anti-MASP1 Rabbit Monoclonal Antibody [clone: EPR22680-36]
Anticuerpos
Rabbit monoclonal [EPR22680-36] to MASP1.
Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MASP1
Clonality: Monoclonal
Clone: EPR22680-36
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat