Anti-SI-CLP Rat Monoclonal Antibody [clone: 1B8]

Proveedor: Abcam
AB305176-20UG AB305176-1MG
ABCAAB305176-20UGEA 244 EUR
ABCAAB305176-20UG ABCAAB305176-1MG
Anti-SI-CLP Rat Monoclonal Antibody [clone: 1B8]
Anticuerpos
Rat monoclonal [1B8] to SI-CLP.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SI-CLP
Clonality: Monoclonal
Clone: 1B8
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR