Anti-TGN46 Rabbit Monoclonal Antibody [clone: EPR24770-16]
Proveedor: Abcam
AB271183-100 AB271183-20UL AB271183-1ML
ABCAAB271183-100EA 713 EUR
ABCAAB271183-100 ABCAAB271183-20UL ABCAAB271183-1ML
Anti-TGN46 Rabbit Monoclonal Antibody [clone: EPR24770-16]
Anticuerpos
Rabbit monoclonal [EPR24770-16] to TGN46.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TGN46
Clonality: Monoclonal
Clone: EPR24770-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human