Anti-Pro-Collagen III alpha 1 Rabbit Monoclonal Antibody [clone: EPR24255-13]

Proveedor: Abcam
AB279338-100 AB279338-20UL AB279338-1ML
ABCAAB279338-100EA 713 EUR
ABCAAB279338-100 ABCAAB279338-20UL ABCAAB279338-1ML
Anti-Pro-Collagen III alpha 1 Rabbit Monoclonal Antibody [clone: EPR24255-13]
Anticuerpos
Rabbit monoclonal [EPR24255-13] to Pro-Collagen III alpha 1.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Pro-Collagen III alpha 1
Clonality: Monoclonal
Clone: EPR24255-13
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR