Anti-beta III Tubulin Mouse Monoclonal Antibody [clone: AA10]

Proveedor: Abcam
AB231083-100 AB231083-1MG
ABCAAB231083-100EA 707 EUR
ABCAAB231083-100 ABCAAB231083-1MG
Anti-beta III Tubulin Mouse Monoclonal Antibody [clone: AA10]
Anticuerpos
Mouse monoclonal [AA10] to beta III Tubulin.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: beta III Tubulin
Clonality: Monoclonal
Clone: AA10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR