Anti-CD137 Rabbit Monoclonal Antibody [clone: EPR17835-39]

Proveedor: Abcam
AB242799-100 AB242799-1
ABCAAB242799-100EA 938 EUR
ABCAAB242799-100 ABCAAB242799-1
Anti-CD137 Rabbit Monoclonal Antibody [clone: EPR17835-39]
Anticuerpos
Rabbit monoclonal [EPR17835-39] to CD137 - BSA and Azide free (Detector).

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD137
Clonality: Monoclonal
Clone: EPR17835-39
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR