Anti-CRISPR-associated endonuclease Cas12a Mouse Monoclonal Antibody [clone: 3D3-F7]

Proveedor: Abcam
AB273438-100 AB273438-1MG AB273438-20UG
ABCAAB273438-100EA 713 EUR
ABCAAB273438-100 ABCAAB273438-1MG ABCAAB273438-20UG
Anti-CRISPR-associated endonuclease Cas12a Mouse Monoclonal Antibody [clone: 3D3-F7]
Anticuerpos
Mouse monoclonal [3D3-F7] to CRISPR-associated endonuclease Cas12a.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CRISPR-associated endonuclease Cas12a
Clonality: Monoclonal
Clone: 3D3-F7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR