Anti-Eph receptor B2 Rabbit Monoclonal Antibody [clone: EPR22427-268]

Proveedor: Abcam
AB252935-100 AB252935-20UL AB252935-1ML
ABCAAB252935-100EA 800 EUR
ABCAAB252935-100 ABCAAB252935-20UL ABCAAB252935-1ML
Anti-Eph receptor B2 Rabbit Monoclonal Antibody [clone: EPR22427-268]
Anticuerpos
Rabbit monoclonal [EPR22427-268] to Eph receptor B2.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Eph receptor B2
Clonality: Monoclonal
Clone: EPR22427-268
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR