Anti-Kappa light chain Rabbit Monoclonal Antibody [clone: EPR23580-16]
Proveedor: Abcam
AB281075-100 AB281075-1
ABCAAB281075-100EA 869 EUR
ABCAAB281075-100 ABCAAB281075-1
Anti-Kappa light chain Rabbit Monoclonal Antibody [clone: EPR23580-16]
Anticuerpos
Rabbit monoclonal [EPR23580-16] to Kappa light chain - BSA and Azide free (Detector).
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Kappa light chain
Clonality: Monoclonal
Clone: EPR23580-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human