Anti-SQSTM1/p62 Mouse Monoclonal Antibody [clone: 3/P62 LCK LIGAND]

Proveedor: Abcam
AB280091-100 AB280091-1
ABCAAB280091-100EA 1020 EUR
ABCAAB280091-100 ABCAAB280091-1
Anti-SQSTM1/p62 Mouse Monoclonal Antibody [clone: 3/P62 LCK LIGAND]
Anticuerpos
Mouse monoclonal [3/P62 LCK LIGAND] to SQSTM1 / p62 - BSA and Azide free.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: SQSTM1 / p62
Clonality: Monoclonal
Clone: 3/P62 LCK LIGAND
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR