Anti-Sall4 Rabbit Monoclonal Antibody [clone: SP289]

Proveedor: Abcam
AB226756-100 AB226756-500 AB226756-1
ABCAAB226756-100EA 700 EUR
ABCAAB226756-100 ABCAAB226756-500 ABCAAB226756-1
Anti-Sall4 Rabbit Monoclonal Antibody [clone: SP289]
Anticuerpos
Rabbit monoclonal [SP289] to Sall4.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Sall4
Clonality: Monoclonal
Clone: SP289
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR