Anti-MUC16 Rabbit Monoclonal Antibody [clone: EPR22287-162]

Proveedor: Abcam
AB274667-100 AB274667-1
ABCAAB274667-100EA 869 EUR
ABCAAB274667-100 ABCAAB274667-1
Anti-MUC16 Rabbit Monoclonal Antibody [clone: EPR22287-162]
Anticuerpos
Rabbit monoclonal [EPR22287-162] to MUC16 - BSA and Azide free (Capture).

Type: Primary
Antigen: MUC16
Clonality: Monoclonal
Clone: EPR22287-162
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR