Anti-eNOS Mouse Monoclonal Antibody [clone: 3/eNOS/NOS Type III]

Proveedor: Abcam
AB282109-100 AB282109-1MG AB282109-20UG
ABCAAB282109-100EA 713 EUR
ABCAAB282109-100 ABCAAB282109-1MG ABCAAB282109-20UG
Anti-eNOS Mouse Monoclonal Antibody [clone: 3/eNOS/NOS Type III]
Anticuerpos
Mouse monoclonal [3/eNOS/NOS Type III] to eNOS.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: eNOS
Clonality: Monoclonal
Clone: 3/eNOS/NOS Type III
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR